Griffin Tubing Glass Cutter
Supplier: Humboldt Manufacturing
H-13520
470122-404EA 32 USD
470122-404
Griffin Tubing Glass Cutter
Cutters
Hardened steel wheel cuts glass tubes up to 11/2" dia. Two V guides align the tubing and wheel is spring loaded to stay open.
  • Made of nickel-plated zinc-alloy die-casting