Regulated Low Voltage DC Power Supply
Supplier: B & K Precision
1665
470122-110EA 773.1 USD
470122-110
Regulated Low Voltage DC Power Supply
Power Supplies
Compact design features excellent regulation and great reliability
This unit allows you to control the voltage and current independent from each other. A digital read out gives the exact value voltage and current so you do not need external meters.