Switching Mode Regulated DC Power Supply
Supplier: B & K Precision
1692
470122-108EA 776.35 USD
470122-108
Switching Mode Regulated DC Power Supply
Power Supplies
Provides high current output in a lightweight and compact package.
  • Lightweight and compact
  • High efficiency
  • Current fold back circuitry with illuminated indicator prevents overloading the power supply
  • Over temperature protection circuitry
  • Over voltage protection prevents abnormal high output voltage
It is suitable for a variety of uses, especially for powering DC operated mobile radio equipment on the bench. It provides a variable voltage output from 3V to 15V at 40A continuous operation. In addition a fixed 13.8 VDC output is also selectable by a slider switch. Switching mode power supplies have the advantage of lightweight and high efficiency when compared to traditional linear mode power supplies. The efficiency can exceed 80% under the best conditions. Advanced circuitry protects against overload and provides immunity from RFI. A bright red and green LED display provides for an accurate and highly readable indicator of settings.