Hanna Instruments pH Electrode Storage Solution (230 ml)
Supplier: HANNA INSTRUMENTS
HI70300M
470122-066EA 15 USD
470122-066
Hanna Instruments pH Electrode Storage Solution (230 ml)
Electrolytes Electrode Storage Solutions
This electrode storage solution prepared with reagent grade chemicals that can be used to ensure optimum performance of your pH and ORP electrodes. Properly storing your pH electrode in a solution keeps the glass membrane well hydrated which maintains proper function and provides accurate readings.
  • Special formulation to minimize bacterial growth on your electrode
  • Designed for storing any pH or ORP electrode
  • Air tight bottle with a tamper-proof seal Ensures quality and freshness of solution
This storage solution is a specially formulated electrode storage solution that can be used to store your pH electrode*. To ensure the best response time, the sensor tip and the reference junction of the pH electrode should be kept moist when not in use. Placing the pH electrode in a small glass filled with storage solution or replacing the solution in the protective cap is a n easy way to store the electrode. This storage solution should also be used to rehydrate the electrode after cleaning by soaking for at least one hour before taking measurements.