Multipurpose Drawing Compass
Supplier: Educational Tools
240-14A-106
470122-024CS 2055.65 USD
470122-024
Multipurpose Drawing Compass
Compasses
Consists of three 2" Ø, joined aluminum disks with knob tightening adjustment which holds pencils, chalk or hobby knives up to 0.6" in Ø. The diameter of the desired circle is set using scale on the front disk.
Ordering information: Supplied with pencil.