Silva Explorer 2.0 Compass
Supplier: LIBERTY MOUNTAIN SPORTS, LLC
544905
470121-828EA 39 USD
470121-828
Silva Explorer 2.0 Compass
Compasses
If you are looking for a basic but yet functional compass for your outdoor adventure, Explorer should be your choice.
  • Detachable lanyard with safety-release
  • Turnable housing usable with the SILVA 1-2-3 System®
  • Declination scale inside the capsule
  • Night use/luminous markings enables night time navigation up to 4 hours once activated by daylight/flashlight
  • Robust material
  • Waterproof
Explorer features DryFlex™ grip for easy handling and comes with a use-anywhere declination scale inside the capsule, a magnifying lens, and base plate map-measuring in inches and scales of 1:62.5k mile and 1:24k USGS scale. The baseplate has a curved-up at the back-end for perfect fit in the hand. The patented red/black North/South lines in the capsule bottom ensures easy and safe settling. The distinct arrow and scales/markings in a clear contemporary font combined with lots of transparency in the baseplate makes navigating easier - when activated in light the luminous markings enable night time navigation. The needle is made from finest Swedish steel with high quality magnetic features.