Day’s Pinchcock
Supplier: Humboldt Manufacturing
H-8630
470121-802EA 5.35 USD
470121-802
Day’s Pinchcock
Clamps General Purpose Clamps
This open-ended pinchcock is made of hardy steel.
  • Nickel-Plated
  • Round Steel Wire Construction
  • Length: 67 mm
  • Pinchcock Takes Tubing of up to 10 mm
It’s open-ended to permit ready attachment and removal without disconnecting the rubber tubing.
The nickel-plated pinchcock is made of round steel wire and is 67 mm long. It takes rubber tubing of up to 10 mm in diameter.