Double Buret Clamp with Vinylized Jaws
Supplier: Wards
CH0258B
470121-774EA 34.3 USD
470121-774
Double Buret Clamp with Vinylized Jaws
Clamps General Purpose Clamps
Grasps any buret firmly from micro to 100 ml size.
Each buret is held independently. Locks to support rod by thumbscrew, which is tightened from the front; will fit support rods up to 13mm (1/2") in diameter.
Vinylized jaws provide nonslip grip and resist attack by acids, alkalies or solvents.