True RMS Bench Digital Multimeter
Supplier: B & K Precision
2831E
470121-628EA 691.99 USD
470121-628
True RMS Bench Digital Multimeter
Electrical Meters Digital Multimeters
A versatile and dependable bench-top digital multimeters suitable for applications.
  • Up to 50,000 count display resolution
  • Basic VDC accuracy of 0.02% for the 5491B and 0.03% for the 2831E
  • Dual display to indicate two measurements simultaneously
  • AC + DC True RMS
  • Up to 25 readings per second measurement rate
Enhance your productivity with built-in math functions and USB connectivity, features not found in other bench meters in this price category.
The meters take typical multimeter measurements such as volts, ohms, and amps with great accuracy and stability and were designed for cost conscious users requiring a basic and dependable meter with a broad range of extra features offered at a value price.