Variable Gap Magnet
Supplier: Wards
PH0801VGM
470121-488EA 97.1 USD
470121-488
Variable Gap Magnet
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Adjust the Magnets According To Your Needs.
  • This apparatus consists of two neodymium magnets set on screws on a metal base
  • Versatile and customizable, the magnet positions can be altered between 3 mm and 83 mm or removed to implement other magnets
  • A pair of metal plates are included to provide a uniform magnetic field when performing experiments