4 Output Power Supply
Supplier: Wards
ESRQP1343
470121-454EA 709.8 USD
470121-454
4 Output Power Supply
Power Supplies
Perfect for driving motors, digital and analog circuits and light sources.
This highly versatile supply comes with digital display for both current and voltages. Housed in high quality plastic case, the unit offers four outputs, with each of the four outputs protected from overload and short circuit.
Certifications: CSA Approved