Hand Pump with Vacuum Plate
Supplier: Wards
PH0166B
470121-430EA 161.9 USD
470121-430
Hand Pump with Vacuum Plate
Pumps Vacuum Pumps
Efficient combination of a pump and a vacuum plate.
  • Plate features a silicon seal, which allows perfect sealing of the bell jar with the plate
  • 20 cm diameter size
Just a few strokes of the pump are enough to establish a vacuum in a bell jar, so students can observe the reducing sound level of the bell. The plate can also be used with electric vacuum pumps with the help of a three-way stopcock under the plate.