Buret Filler, Eisco Scientific
Supplier: Wards
BTAUT50
470121-260EA 82 USD
470121-260
Buret Filler, Eisco Scientific
Burets
Automatic 50ml burette made of borosilicate glass which is self-zeroing
0.1ml resolution with permanent blue markings against a white background for easy viewing.  Stopcock is pushbutton style for ease of use to deliver fluids drop by drop.   Attached to the stop cock is a screw clamp for repeat titrations.  This set up is a complete self-supporting structure with a heavy duty base made of polyethylene which the reservoir snaps into.  The advantage to a system such as this is that it is a completely closed system.