CENCO® Large Forced Oscillation and Resonance Demonstrator
Supplier: VWR International
470121-166
470121-166EA 1071.6 USD
470121-166
CENCO® Large Forced Oscillation and Resonance Demonstrator
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Study Static and Dynamic Oscillations
Study Static and Dynamic Oscillations
This oversized apparatus is suitable for demonstrations in large classrooms, so all students can observe. Apply weights and note the spring elongations along the graduated scale. An acrylic glass cylinder, filled with oil or water, is used for study damping. The friction ratio can also be varied through changing discs. Excitation frequency is displayed on the LCD panel. Requires low voltage power supply available separately. Size: 35cm L x 35cm W x 85cm H.