SCIENCEWARE® Vacuum Aspirator Bottle
Supplier: Bel-Art
F19917-0001_SE
470121-056EA 349.55 USD
470121-056
SCIENCEWARE® Vacuum Aspirator Bottle
Bottles Aspirators
For easy and continuous vacuum aspiration of a variety of liquids including tissue culture media, wash solutions, supernatant fluids after centrifugation, and chemical waste.
  • Heavy-walled design
  • Polypropylene construction
  • Housed in an epoxy-coated steel wire cage
  • Flexible collection tubing
Consists of a heavy-walled polypropylene collection bottle housed in an epoxy-coated steel wire cage, a secondary vacuum trap, and flexible collection tubing. The collection bottle safely retains fluid while the attached trap protects the vacuum source from accidental fluid backflow. The flexible collection tubing accepts Pasteur pipets, regular pipets, or pipettor tips approximately 0.70 mm in diameter for work with Petri dishes, microtiter plates, flasks, and centrifuge tubes. The small footprint of the wire cage supports the collection bottle and the secondary vacuum trap without overcrowding the work space.
Bottle and cap are autoclavable.