Frigimat® Cub Dry Ice Maker
Supplier: Bel-Art
F38874-0000_SE
470121-042EA 736.8 USD
470121-042
Frigimat® Cub Dry Ice Maker
Ice Makers
Attach this compact unit to a liquid CO2 cylinder and in a few minutes you’ll have a ready-to-use 250 - 350 g block of dry ice.
  • Dry ice production can be viewed through a translucent polycarbonate body that opens without clamps or screws
  • The system operates multiple pressure relief safeguards
Weight: 4.8 lbs.