Specific Gravity Bottles
Supplier: American Scientific
3505-02
470120-780EA 16.7 USD
470120-780
Specific Gravity Bottles
Specific Gravity Bottles
Also called density bottles or R.D. bottles, each has a perforated stopper (about 1 mm bore) and a ground mouth for proper fit. Made of Borosilicate glass. Uncalibrated.