Dissectible Coils and Cores
Supplier: Wards
PH0850CSET
470120-754EA 396.9 USD
470120-754
Dissectible Coils and Cores
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
This set features five coils.
  • Coils Illustrate Electromagnetism and Induction
  • Useful in Basic Transformer Experiments
  • Included Coils Have 200, 400, 800, 1600, and 3200 Turns
  • Five Coils in Set
The coils illustrate electromagnetism and induction. They are also useful in basic transformer experiments.
The included coils have 200, 400, 800, 1600, and 3200 turns.