U-Form Electromagnet
Supplier: Wards
PH0800B
470120-696EA 29.76 USD
470120-696
U-Form Electromagnet
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Quality electromagnet.
  • Wound with insulated copper wire
  • Provided with barrel connections
This unique electromagnet consists of an iron U-shaped core with armature and a carrying hook. The electromagnet is designed for use on 4–6 volts DC.