Steam Engine Model
Supplier: Wards
PH0483
470120-694EA 143 USD
470120-694
Steam Engine Model
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Demonstrate Principles of the First Commercially Produced Steam Engine.
No longer in frequent use, the steam engine was one of the first inventions which sparked an industrial revolution. This all metallic model enables you to show how this once revolutionary engine works well still today. The metal cylinder is section cut and covered with a glass plate for easy viewing within. Working parts include a piston and slide valve, so link motion and reversing method can be observed. The movement of the wheel can even be demonstrated by blowing on it.