Forced Oscillation and Resonance Demonstrator
Supplier: VWR International
PH0753FOR
470120-686EA 146.2 USD
470120-686
Forced Oscillation and Resonance Demonstrator
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Simple Mechanical System Clearly Conveys Complex Physics Principles.
  • Includes an instruction manual
Designed to show the vibration of an object when repeated impulses are applied, this demonstrator consists of two weights of different natural frequencies, two cams driven by a 6V motor with speed control, and four springs. The speed control is used to adjust the frequency of applied force for resonance, so a greater degree of variance is possible.