VWR® Volumetric Flasks, Class B
Supplier: VWR International
470120-670 470120-672 470120-674 470120-676 470120-678 470191-248
470120-670PK 372.03 USD
470120-670 470120-672 470120-674 470120-676 470120-678 470191-248
VWR® Volumetric Flasks, Class B
Flasks Volumetric Flasks
Our new high-quality borosilicate volumetric flasks meet ASTM standards and are a great value.
  • Borosilicate glass construction
  • Snap-on caps
  • Graduated line for accurate measurements
Class B with capacity tolerances twice those specified for Class A volumetric work. The necks are tooled to fit the included snap-caps. With graduation line for accurate measurements.