Vernier® Ammonium Ion-Selective Electrode
Supplier: Vernier Software & Technology
NH4-BTA
470120-632EA 313.99 USD
470120-632
Vernier® Ammonium Ion-Selective Electrode
Electrodes ISE Electrodes
Measure concentration of ammonium in aqueous samples.
  • Combination-style, non-refillable, gel-filled electrode.
Use the Ammonium ISE to measure levels of ammonium ions introduced from fertilizers. It can also indicate aqueous ammonia levels if sample solutions are acidified to convert NH3 to NH4+.
The Ammonium ISE has a combination-style, non-refillable, gel-filled electrode. Like all other PVC ISE membranes, the membrane on the ISE has a limited life expectancy. However, the replaceable module of ISE allows you to simply discard the used membrane module, and replace it with a new one.
Note: We recommend that you do not purchase ISE replacement modules too far in advance of their expected time of use as degradation occurs while they are stored on the shelf.
Important: Ion-Selective Electrodes require good chemical technique and careful calibration to obtain accurate results; they are not recommended for middle school or elementary students.
For educational use only.