Micro Burner with Wide Tube for Artificial Gases
Supplier: Humboldt Manufacturing
H-5805
470120-590EA 36.95 USD
470120-590
Micro Burner with Wide Tube for Artificial Gases
Heaters Gas Burners
Use with artificial gas with a heat value from 300 to 800 BTU. With nickel-plated die-cast zinc alloy base, die-cast nipple, aluminum ¹/₄" mixing tube, safety pilot ring and adjustable air ports.
  • For 6 mm Int.Ø tubing
  • Height: 3¹/₃", mixing tube dia. ⁷/₁₆"