Glass Buret Funnell, Montclare Scientific
Supplier: Montclare
QUOTE2-93
470120-586EA 31.6 USD
470120-586
Glass Buret Funnell, Montclare Scientific
Funnels Filtration Funnels Filter Funnels
For filling burets. Diameter: 35 mm; total length: 60 mm; outside diameter of stem: 6 mm.