Laerdal® Infant I.O. Training Leg
Supplier: American 3B Scientific
1005240
470120-404EA 563.05 USD
470120-404
Laerdal® Infant I.O. Training Leg
Educational Materials Biology Educational Materials Human Biology Learning Activities
Use Alone, Or As Part Of The Nursing Baby Sim
  • Simulated tibia and anatomical landmarks at the tibial tuberosity and medial malleolus
  • Fluid may be infused for realistic flashback
  • Drain in heel connects to reservoir bag
Lifelike reproduction of infant lower leg allows students to master infant intraosseous infusion techniques. Simulated tibia and anatomical landmarks at the tibial tuberosity and medial malleolus. Fluid may be infused for realistic flashback. Drain in heel connects to reservoir bag.