Laerdal® Infant I.V. Training Arm
Supplier: American 3B Scientific
1005239
470120-402EA 538.59 USD
470120-402
Laerdal® Infant I.V. Training Arm
Educational Materials Biology Educational Materials Human Biology Learning Activities
Use Alone, Or As Part Of The Nursing Baby Sim
  • Venipuncture possible in antecubital fossa and dorsum of hand
  • Simulated blood may be infused for realistic flashback
Lifelike reproduction of an infant arm for mastering extremity venipuncture procedures and fluid administration in the superficial veins of the hand. Phlebotomy and Venipuncture are possible in antecubital fossa and dorsum of the hand. Fluid may be infused for realistic flashback.