Laerdal® Nursing Kid Manikin
Supplier: American 3B Scientific
1005244 1005275
470120-378EA 6787.8 USD
470120-378 470103-376 470103-392
Laerdal® Nursing Kid Manikin
Models Human Biology Models
Clinical Training On A 6 Year Old Child Trainer
  • Highly mobile for use in multiple clinical settings and hospital transport
  • Efficient training care and management of a variety of both common and uncommon in-hospital pediatric patients including wound assessment and care, first aid and child abuse training
  • Durable, rugged and lifelike; made to withstand years of use
The clinical skills training manikin for in-hospital pediatric professionals. Nursing Kid is a training manikin realistically representing a six-year old child. He is designed for skill and scenario based training of the care and management of a variety of pediatric in-hospital patients. Training includes sound auscultation, IV cannulation, urinary catheterization, and general pediatric patient care. This child is ideal for clinical training in core pediatric in-hospital clinical skills. Features: Head with anatomical landmarks, trachea, and esophagus along with simulated lungs and stomach allow the practice of many procedures, including NG, OG, tracheal care and suctioning, and insertion, securing and care of endotracheal tube. Allows for the auscultation and diagnosis of normal and abnormal hear, breath and bowel sounds. Bilateral deltoid, bilateral thigh, gluteal intramuscular injections possible. Articulating IV arm allows for practice of IV cannulation, medication administration and site care and maintenance. Presents normal and abnormal heart, breath and bowel sounds for auscultation. Supplied with: Full-Body Pediatric Manikin, manikin lubricant, simulated blood, Hospital Gown, Carry Case, Directions for Use.
Ordering information: Does not include a SimPad Unit.