3B Scientific® MICROanatomy™ Eye Model
Supplier: American 3B Scientific
1000260
470120-372EA 572.99 USD
470120-372
3B Scientific® MICROanatomy™ Eye Model
Models Human Biology Models
A Histologic view of the eye.
  • Three-Year Manufacturer's Warranty
  • Incredible Detail and Enlargement
  • Microscopic Structure of Photoreceptors and the Cells of the Pigmented Layer
  • Includes 3B Smart Anatomy
The left side of the model shows the complete structure of the retina including the supplying vascular layer and parts of the sclera. The right part of the eye model is a sectional enlargement.
This model shows the microscopic structure of the photoreceptors and the cells of the pigmented layer.
Accessories information: For information about 3BMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable