ADC® Diagnostix 703 Palm Aneroid Sphygmomanometers
Supplier: AMERICAN DIAGNOSTIC CORP CP
703-7IBK W51467 W51471 W51472
470119-700CS 2488.05 USD
470119-700 470119-702 470119-708 470119-710
ADC® Diagnostix 703 Palm Aneroid Sphygmomanometers
Blood Pressure Monitors
An easy to use aneroid sphyg.
  • Palm style manometer with integrated bulb
  • Bayonet connector for easy cuff changes
  • Not made with natural rubber latex
Palm aneroid sphygmomanometers are favored by those who need to rapidly take blood pressure, and may need to quickly change cuffs. The Diagnostix 703 is a great example of this type of sphygmomanometer. The manometer is a precision crafted movement, contained in a zinc cage. The cage is further protected by the durable polymer housing. An inflation bulb, and deflation valve are also contained in the housing. Deflation is easily accomplished with a single finger using the ambidextrous trigger. The included cuff attaches to the unit with a single tube using a bayonet mount for fast and accurate changes. The ADC cuff is made of high quality nylon with an internal bladder.
703 Series: Versatile palm style aneroid sphygmomanometer, that combines precision crafted performance with rugged durability. Integrated bulb, gauge and release for ambidextrous one-hand convenience.