ADC® Pocket Ophthalmoscope Set
Supplier: AMERICAN DIAGNOSTIC CORP CP
5112N
470119-692CS 1110 USD
470119-692
ADC® Pocket Ophthalmoscope Set
Opthalmoscopes
This pocket ophthalmoscope set features a 2.5 V halogen light source interchangeable “AA” metal battery handle.
  • Impact resistant head
Pocket clip with integral on/off slide switch for ambidextrous use. Lifetime quality performance guarantee on optics.