Tactile Monofilament Evaluator, Foot Set
Supplier: American 3B Scientific
3005262
470119-636EA 154.7 USD
470119-636
Tactile Monofilament Evaluator, Foot Set
Clinical Diagnostic Supplies Clinical Instruments
The baseline tactile monofilament evaluator, foot set, measures cutaneous sensory perception threshold of the feet. Used to test sensation levels, and obtain objective data for accurate reporting on the status of diminishing or retuning sensibility.
Features 2.83, 3.61, 4.31, 4.56, 5.07 and 6.65 sized monofilaments.