Baseline® Tactile Monofilaments Sensory Evaluators
Supplier: American 3B Scientific
3005261
470119-634EA 512.35 USD
470119-634
Baseline® Tactile Monofilaments Sensory Evaluators
Clinical Diagnostic Supplies Clinical Instruments
Quantify Sensation Levels.
  • Units measure cutaneous sensory perception thresholds
  • Each monofilament represents a unique amount of force
  • Twenty monofilaments included
  • Protective case included
These evaluators help test sensation levels and obtain objective data for accurate reporting on the status of diminishing or retuning sensibility. They measure cutaneous sensory perception thresholds. Each monofilament represents a unique amount of force ranging from 1.65 to 6.65 in size and .008 to 300 grams of force. The full set stores in a protective case.