Pinch Gauge, AMERICAN 3B SCIENTIFIC SE
Supplier: American 3B Scientific
1009010
470119-598EA 329 USD
470119-598
Pinch Gauge, AMERICAN 3B SCIENTIFIC SE
Dynamometers
This 6.35 cm (2,5") diameter head hydraulic pinch gauge offers accurate and repeatable pinch strength measurements. The hydraulic system assures convenience, product reliability, measurement accuracy and repeatability. For all pinch test: Tip, key and palmar.
Registers up to 50 lbs. (22.7 kg)