Hand Evaluation Set, Three Pieces, AMERICAN 3B SCIENTIFIC SE
Supplier: American 3B Scientific
1009008
470119-594EA 849 USD
470119-594
Hand Evaluation Set, Three Pieces, AMERICAN 3B SCIENTIFIC SE
Dynamometers
The baseline hydraulic hand evaluation set has all instrument necessary to evaluate strength and range of motion of hand and fingers. The hydraulic system assures convenience, product reliability, measurement accuracy and repeatability.
  • Standard head hand dynamometer is used by therapists
  • The standard 6.35 cm (2.5") diameter head is used throughout the industry as most popular size
Delivery information: Includes a 200 lbs. (90 kg) hydraulic hand dynamometer, 50 lbs. (22.6 kg) hydraulic pinch gauge and stainless steel finger goniometer.