Baseline® Hand Evaluation Set, Seven Pieces
Supplier: American 3B Scientific
1009007
470119-592EA 899 USD
470119-592
Baseline® Hand Evaluation Set, Seven Pieces
Educational Materials Biology Educational Materials Physiology Learning Activities
A full analog evaluation set.
  • Seven piece evaluation set
  • Includes case for storage and transport
The Baseline hand evaluation set includes all instruments needed for a full hand evaluation.
Grip strength is measured by the 200 lbs. hydraulic hand dynamometer and pinch strength by the mechanical 30 lbs. pinch gauge.
Delivery information: Includes a stainless steel finger goniometer 5 cm (2"), two-point discriminator with third point, Wartenburg pinwheel, finger circumference gauge, and functional finger motion gauge in a hard shell carrying case.