Back-Leg-Chest Dynamometer, AMERICAN 3B SCIENTIFIC SE
Supplier: American 3B Scientific
1015293
470119-590EA 697.8 USD
470119-590
Back-Leg-Chest Dynamometer, AMERICAN 3B SCIENTIFIC SE
Dynamometers
Back-leg-chest dynamometer's to measure the strength of a patients back, leg or chest. Oversized body includes a solid base for safety, as well as cushion handgrips for comfort. Chain adjusts for height differences or to vary the point of force application.
Strength indicator remains at subject's maximum reading until reset.
Scale measure to 300 kg (660 lbs.).