Skinfold Caliper, 3B Scientific
Supplier: American 3B Scientific
1009006
470119-588EA 256.7 USD
470119-588
Skinfold Caliper, 3B Scientific
Calipers
This lightweight aluminum skinfold caliper includes a measurement arm with "floating tips" that under constant pressure assures accurate and reproducible results.
Easy to read 60 mm scale.
Includes booklet, tables and caliper carrying case.