Digital Inclinometer
Supplier: American 3B Scientific
3005247
470119-586EA 199 USD
470119-586
Digital Inclinometer
Clinometers
Motion in its total range can be read from the LED panel, after the joint has been taken through the digital inclinometer. Place the inclinometer near the joint to be measured.
Press "zero" button. Move joint through its range. "HOLD" button.
Includes carrying case.