Arrhythmia, Heart and Breath Sounds Tutor
Supplier: American 3B Scientific
1018150 1005892 1018181 1018182 1018183 1018184 1018185 1018186 1018188 1018189 1018190 1018191 1018192 1005891 1018193 1018194 1018175 1018176 1018177 1018178 1018179 1018180
470119-490EA 2103.15 USD
470119-490 470119-492 470119-494 470119-496 470119-498 470119-500 470119-502 470119-504 470119-506 470119-510 470119-512 470119-514 470119-516 470119-518 470119-520 470119-522 470119-524 470119-526 470119-528 470119-530 470119-532 470119-534 470119-536
Arrhythmia, Heart and Breath Sounds Tutor
Models Human Biology Models
Flexible And Adaptable Training
  • Multiple cartridges make it easy to change sounds
  • Students all hear the same sounds for ease of identification
Combining two tutors into one, this unit simulates either heart and breath sounds or arrhythmias. A perfect tool for Medical Schools, Nursing schools, EMS Education programs or other healthcare settings where rhythm and sound recognition is critical. This unique tutor allows instructors the flexibility to change menus by changing plug-in modules. The unit has a 16-panel keypad with two built-in sounders and two external sounders. Linking additional sounders or a speaker allows access to more users. There is a volume control and two LEDs that illuminate during systole as well as an illuminated display showing the example currently playing. Arrhythmias may be viewed by connecting the three snap-on lead patient wires to your ECG-monitor. The unit comes with a 12 Volt adapter. All sound menus are sold separately, each containing 16 different sounds.
All data used in the menus are actual patient sounds. Arrhythmias and sounds of the same name are from different patients to show appearance variations.