Venous Access Device Model
Supplier: American 3B Scientific
1005563
470119-452EA 1291.29 USD
470119-452
Venous Access Device Model
Models Human Biology Models
Inexpensive Catheter Cleaning Training
  • Practice cleaning 2 catheter systems
  • Viewing port to access one of the implanted devices
  • Includes carrying case
Teach patients and healthcare staff the care and use of two different types of catheters with this model. Features left and right subclavian catheter connections that may be flushed with water; implanted venous access system with a "skin" flap over the implant to allow viewing. Made of skinned, lightweight urethane foam, this model may be washed with soap and water, or painted with antiseptics.
Mounted on a collapsible easel and packed in its own carrying case.