Life/form® Bandaging Simulator With Ostomy
Supplier: Nasco
LF00929
470119-390EA 1664.85 USD
470119-390
Life/form® Bandaging Simulator With Ostomy
Models Human Biology Models
Just Like A Patient, But Doesn'T Wriggle.
  • Practice bandaging 14 different wounds
  • Reformulated skin allows for better dressing adhesion
  • Includes carrying case
This simulator consists of a female torso with flexible, lifelike skin which realistically responds to adhesives and all types of bandaging procedures. 14 different wounds allow one to practice wound management techniques, cleaning and bandaging techniques just like on a real patient. Wound closures feature surgical staples and sutures which cannot be removed.