Laerdal® Baby Hippy Trainer
Supplier: American 3B Scientific
1005864
470119-308EA 1215.09 USD
470119-308
Laerdal® Baby Hippy Trainer
Models Human Biology Models
Neonatal Skills
  • Practice hip setting and assessment procedures
  • Natural feel simulator
Representation of the lower torso and limbs of a newborn female with: Dislocated left hip for practice of the Ortolani Jerk-Sign and Lax right hip for performing the Barlow maneuver.