Simulaids® Gynecologic Training
Supplier: Nasco
160-1900
470119-278EA 1409.85 USD
470119-278
Simulaids® Gynecologic Training
Models Human Biology Models
Basic Skills And Diagnostic Practice
  • Easy to exchange parts allow for multiple scenarios
  • Includes 5 cervixes and 3 uteri
A lifelike female pelvis for developing diagnostic skills in gynecologic procedures, anatomical instruction, abdominal palpation, and speculum instruction. EVA allows the comprehensive, stress free introduction of gynecological examinations, or more advanced tactile comparisons of pelvic pathologic conditions in a classroom setting easing the way into clinical experience. Constructed of soft, easily cleaned, vinyl materials that approximate skin texture, EVA provides detailed, life-like anatomical features and references. Included are the following interchangeable inserts: normal cervix for IUD insertion and removal, normal parous cervix, cervix with endocervical polyp, cervix with ectropion characteristics, cervix with neoplasia (carcinoma), normal uterus for IUD insertion and removal, pregnant 10-week uterus, and 2 adnexal masses.