Life/form® Birthing Station Simulator
Supplier: Nasco
LF01070
470119-244EA 2281 USD
470119-244
Life/form® Birthing Station Simulator
Models Human Biology Models
Learn Palpation Techniques
  • Three head positions in relation to ischial spines
  • Includes carrying case
This birthing simulator is especially designed to demonstrate and practice the palpation of the birth canal. It shows the relationship between fetal head and ischial spines of the pelvic bone during birth. A simulated fetal head of the birth simulator can be locked in three different stations to demonstrate its position above, between and below the ischial spines. This anatomical birthing model is a great addition to any lesson about the human birth process. The birthing simulator is delivered with storage case. It is great for any gynecological and birth training.