3B Scientific® Tongue And Mouth Floor
Supplier: American 3B Scientific
1002502
470119-096EA 433.35 USD
470119-096
3B Scientific® Tongue And Mouth Floor
Models Human Biology Models
Large Size for Easy Examination.
  • Three-Year Manufacturer's Warranty
  • Removable Base
This tongue model shows the lower jaw up to the second molar and the tongue with mouth-floor musculature in median section and the right sublingual and submandibular gland.