3B Scientific® Classic Tooth Models
Supplier: American 3B Scientific
1017588 1000240 1000241 1000242 1000243 1017580
470119-082EA 773.1 USD
470119-082 470119-084 470119-086 470119-088 470119-090 470119-092
3B Scientific® Classic Tooth Models
Models Human Biology Models
Clearly See Details
  • Includes 3B Smart Anatomy
A classic tooth series showing 5 representative types of adult dentition. Individually mounted on removable stands to display: 2-part lower incisor with longitudinal section, 2-part lower canine with longitudinal section, lower single-root pre-molar, 2-part lower twin-root molar with longitudinal section showing caries attack and 3-part upper triple-root molar with longitudinal section and caries insert. Purchase individually, or as a set of all 5
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.