3B Scientific® Brain Ventricle
Supplier: American 3B Scientific
1001262
470119-062EA 279.7 USD
470119-062
3B Scientific® Brain Ventricle
Models Human Biology Models
This model shows both side ventricles of the brain, the third and fourth ventricle and the Aquaeductus cerebri (Sylvius).
  • 5X ife size
  • 3 year manufacturers warranty
  • Includes 3B smart anatomy
The brain ventricles are life-size. This brain ventricle model is delivered on stand.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.