3B Scientific® Deluxe Head
Supplier: American 3B Scientific
1000217
470119-052EA 810 USD
470119-052
3B Scientific® Deluxe Head
Models Human Biology Models
This six-part head is mounted on a base and features a removable four-part brain half with arteries.
  • 3 year manufacturers warranty
  • Includes 3B smart anatomy
The eyeball with optic nerve is also removable and one side of the high quality model exposes the nose, mouth cavity, pharynx, occiput and skull base. On a removable base for easy display in a classroom or doctor's office.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.