3B Scientific® Skin Cancer Model
Supplier: American 3B Scientific
1000293
470119-038EA 126.05 USD
470119-038
3B Scientific® Skin Cancer Model
Models Human Biology Models
This 3B Scientific® Skin Pathology model shows healthy skin and 5 different stages of malignant melanoma on the front and back, enlarged 8 times. In the top view of the skin cancer model, the individual stages of externally visible skin changes are shown, allowing for an assessment according to the 'ABCDE' criteria.
  • Magnetic assembly
  • Shows micro-anatomical structure from different parts of the body
  • 3 year manufacturers warranty
  • Includes 3B smart anatomy
The sides of the skin cancer model show the various levels of invasion into the skin layers according to Clark (I-V) and the tumor thickness according to Breslow (in mm). 5 original color illustrations on the base of the skin cancer model show various types of malignant melanomas. The skin cancer model comes mounted on a base.
Accessories information: For information about 3B smart anatomy, please see the document attached on this page.